Learn More
MSI1, Mouse, Clone: 2A12, Abnova™
Mouse monoclonal antibody raised against partial recombinant MSI1.
Brand: Abnova MAB10814.100uL
Description
This gene encodes a protein containing two conserved tandem RNA recognition motifs. Similar proteins in other species function as RNA-binding proteins and play central roles in posttranscriptional gene regulation. Expression of this gene has been correlated with the grade of the malignancy and proliferative activity in gliomas and melanomas. A pseudogene for this gene is located on chromosome 11q13. [provided by RefSeq
Specifications
| MSI1 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Flow cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. | |
| MSI1 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Flow Cytometry, Western Blot | |
| 2A12 | |
| Mouse monoclonal antibody raised against partial recombinant MSI1. | |
| In ascites (0.03% sodium azide) | |
| MSI1 | |
| Recombinant protein corresponding to human MSI1. | |
| RUO | |
| 4440 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.