missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRRF, Mouse, Clone: AT7D10, Abnova™
Mouse monoclonal antibody raised against partial recombinant MRRF.
Brand: Abnova MAB11203.100uL
This item is not returnable.
View return policy
Specifications
| MRRF | |
| Monoclonal | |
| 1 mg/mL | |
| Mouse monoclonal antibody raised against partial recombinant MRRF. | |
| In Phosphate-Buffered Saline (pH 7.4) containing 10% Glycerol, 0.01% Sodium azide | |
| MRFF/MTRRF/RRF | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 92399 | |
| Store at 4°C. For long term storage aliquot and store at -20°C. Avoid repeated freezing and thawing cycles. | |
| IgG1 κ |
| ELISA, Western Blot | |
| AT7D10 | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| MRRF | |
| MRRF | |
| Recombinant protein corresponding to amino acids 56-262 of human MRRF. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction