Learn More
MPO, Mouse, Clone: 9B12G7, 4D8B12, 9B12D9, 9C11A5, Abnova™
Mouse monoclonal antibody raised against partial recombinant MPO.
Brand: Abnova MAB10590.100uL
Description
Myeloperoxidase (MPO) is a heme protein synthesized during myeloid differentiation that constitutes the major component of neutrophil azurophilic granules. Produced as a single chain precursor, myeloperoxidase is subsequently cleaved into a light and heavy chain. The mature myeloperoxidase is a tetramer composed of 2 light chains and 2 heavy chains. This enzyme produces hypohalous acids central to the microbicidal activity of netrophils. [provided by RefSeq
Specifications
| MPO | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
| MPO | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 4D8B12, 9B12D9, 9B12G7, 9C11A5 | |
| Mouse monoclonal antibody raised against partial recombinant MPO. | |
| In ascites (0.03% sodium azide) | |
| MPO | |
| Recombinant protein corresponding to amino acids 1-193 of human MPO. | |
| RUO | |
| 4353 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.