missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse anti-Monkey IgA Secondary Antibody
Description
This antibody recognizes monkey IgA and does not cross-react with IgG or IgM; it has been shown to detect secreted IgA in cat saliva and rhesus monkey jejunal washes.
Anti-Monkey secondary antibodies are affinity-purified antibodies with well-characterized specificity for non-human primate (e.g. monkey) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Specifications
| Antigen | Monkey IgA |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | IgA5-3B |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with <0.1% sodium azide |
| Host Species | Mouse |
| Immunogen | purified IgA from serum. |
| Purification Method | Protein A |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?