Learn More
MEIS2, Mouse, Clone: 1B3, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant MEIS2.
Brand: Abnova H00004212-M02A.200uL
Description
This gene encodes a homeobox protein belonging to the TALE ('three amino acid loop extension') family of homeodomain-containing proteins. TALE homeobox proteins are highly conserved transcription regulators, and several members have been shown to be essential contributors to developmental programs. Multiple transcript variants encoding distinct isoforms have been described for this gene. [provided by RefSeq]
Sequence: MDGVGVPASMYGDPHAPRPIPPVHHLNHGPPLHATQHYGAHAPHPNVMPASMGSAVNDALKRDKDAIYGHPLFPLLALVFEKCELATCTPREPGVAGGDVCSSDSFNEDIAVFAKQVRAEKPLFSSNPELDNLMIQAIQVLRFHLLELEKVHELCDNFCHRYISCLKGKMPIDLVIDERDGSSKSDHEELSGSSTNLADHNPSSWRDHDDATSTHSAGTPGPSSGGHASQSGDNSSEQGDGLDNSVASPGTGDDDDPDKDKKRQKKRGIFPKVATNIMRAWLFQHLTHPYPSEEQKKQLAQDTGLTILQVNNWFINARRRIVQPMIDQSNRAVSQGAAYSPEGQPMGSFVLDGQQHMGIRPAGPMSGMGMNMGMDGQWHYMSpecifications
| MEIS2 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| BC001516 | |
| MEIS2 | |
| MEIS2 (AAH01516,1 a.a. ∽ 381 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 4212 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 1B3 | |
| Mouse monoclonal antibody raised against a full-length recombinant MEIS2. | |
| MEIS2 | |
| HsT18361/MGC2820/MRG1 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.