Learn More
MEF2B, Mouse, Clone: 4B5, Abnova™
Mouse monoclonal antibody raised against a partial recombinant MEF2B.
Brand: Abnova H00004207-M24A.200uL
Description
This gene represents numerous read-through transcripts that span geneID:729991 and 100271849. Many read-through transcripts are predicted to be nonsense-mediated decay (NMD) candidates, and are thought to be noncoding. Some transcripts are predicted to be capable of translation reinitation at a downstream AUG, resulting in expression of at least one isoform of myocyte enhancer factor 2B (MEF2B) from this read-through locus. At least one additional MEF2B variant and isoform can be expressed from a downstream promoter, and is annotated on geneID:100271849. [provided by RefSeq
Sequence: SRPSPFRPAAPKAGPPGLVHPLFSPSHLTSKTPPPLYLPTEGRRSDLPGGLAGPRGGLNTSRSLYSGLQNPSpecifications
| MEF2B | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_005919 | |
| MEF2B | |
| MEF2B (NP_005910.1,165 a.a. ∽ 235 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 4207 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 4B5 | |
| Mouse monoclonal antibody raised against a partial recombinant MEF2B. | |
| MEF2B | |
| FLJ32599/FLJ46391/MGC189732/MGC189763/RSRFR2 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.