Learn More
MDM4, Mouse, Clone: 2D10F4, Abnova™
Mouse monoclonal antibody raised against recombinant MDM4.
Brand: Abnova MAB10353.100uL
Beskrivning
The human MDM4 gene, which plays a role in apoptosis, encodes a 490-amino acid protein containing a RING finger domain and a putative nuclear localization signal. The MDM4 putative nuclear localization signal, which all Mdm proteins contain, is located in the C-terminal region of the protein. The mRNA is expressed at a high level in thymus and at lower levels in all other tissues tested. MDM4 protein produced by in vitro translation interacts with p53 via a binding domain located in the N-terminal region of the MDM4 protein. MDM4 shows significant structural similarity to p53-binding protein MDM2. Two transcript variants, one protein-coding and the other likely not to be protein-coding, have been found for this gene. [provided by RefSeq
Specifikationer
| MDM4 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Immunofluorescence (1:200-1:1000) The optimal working dilution should be determined by the end user. | |
| MDM4 | |
| MDM4 | |
| Recombinant protein corresponding to human MDM4. | |
| RUO | |
| 4194 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 2D10F4 | |
| Mouse monoclonal antibody raised against recombinant MDM4. | |
| In ascites (0.03% sodium azide) | |
| DKFZp781B1423/HDMX/MDMX/MGC132766/MRP1 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.