Learn More
MDH1, Mouse, Clone: 99n65, Abnova™
Mouse monoclonal antibody raised against full-length recombinant MDH1.
Brand: Abnova MAB8873.100uL
Description
Malate dehydrogenase catalyzes the reversible oxidation of malate to oxaloacetate, utilizing the NAD/NADH cofactor system in the citric acid cycle. The protein encoded by this gene is localized to the cytoplasm and may play pivotal roles in the malate-aspartate shuttle that operates in the metabolic coordination between cytosol and mitochondria. [provided by RefSeq
Specifications
| MDH1 | |
| Monoclonal | |
| 1 ug/uL | |
| Mouse monoclonal antibody raised against full length recombinant MDH1. | |
| In citrate-Tris-HCl, pH7.0 (0.02% Proclin 300) | |
| MDH-s/MDHA/MGC:1375/MOR2 | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 4190 | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 99n65 | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| MDH1 | |
| MDH1 | |
| Recombinant protein corresponding to full length of human MDH1. | |
| 100 μL | |
| Primary | |
| This antibody recognizes recombinant protein MDH1. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.