Learn More
MDFI, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant MDFI.
Brand: Abnova H00004188-A01.50uL
Description
This protein is a transcription factor that negatively regulates other myogenic family proteins. Studies of the mouse homolog, I-mf, show that it interferes with myogenic factor function by masking nuclear localization signals and preventing DNA binding. Knockout mouse studies show defects in the formation of vertebrae and ribs that also involve cartilage formation in these structures. [provided by RefSeq
Sequence: PGLEVVTGSTHPAEAAPEEGSLEEAATPMPQGNGPGIPQGLDSTDLDVPTEAVTCQPQGNPLGCTPLLPNDSGHPSELSpecifications
| MDFI | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant MDFI. | |
| MDFI | |
| I-MF/I-mfa | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_005586 | |
| MDFI | |
| MDFI (NP_005577, 33 a.a. ∽ 110 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 4188 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only