Learn More
MCM2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant MCM2.
Brand: Abnova H00004171-A01.50uL
Description
The protein encoded by this gene is one of the highly conserved mini-chromosome maintenance proteins (MCM) that are involved in the initiation of eukaryotic genome replication. The hexameric protein complex formed by MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. This protein forms a complex with MCM4, 6, and 7, and has been shown to regulate the helicase activity of the complex. This protein is phosphorylated, and thus regulated by, protein kinases CDC2 and CDC7. [provided by RefSeq
Sequence: TQKFSVMRSMRKTFARYLSFRRDNNELLLFILKQLVAEQVTYQRNRFGAQQDTIEVPEKDLVDKARQINIHNLSAFYDSELFRMNKFSHDLKRKMILQQFSpecifications
| MCM2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant MCM2. | |
| MCM2 | |
| BM28/CCNL1/CDCL1/D3S3194/KIAA0030/MGC10606/MITOTIN/cdc19 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| BC007670 | |
| MCM2 | |
| MCM2 (AAH07670, 805 a.a. ∽ 904 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 4171 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only