missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBNL1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length recombinant MBNL1.
Brand: Abnova H00004154-A01.50uL
This item is not returnable.
View return policy
Description
Sequence: MAVSVTPIRDTKWLTLEVCREFQRGTCSRPDTECKFAHPSKSCQVENGRVIACFDSLKGRCSRENCKYLHPPPHLKTQLEINGRNNLIQQKNMAMLAQQMQLANAMMPGAPLQPVPMFSVAPSLATNASAAAFNPYLGPVSPSLVPAEILPTAPMLVTGNPGVPVPAAAAAAAQKLIRTDRLEVCREYQRGNCNRGENDCRFAHPADSTMIDTNDNTVTVCMDYIKGRCSREKCKYFHPPAHLQAKIKAAQYQVNQAAAAQAAATAAAMGIPQAVLPPLPKRPALEKTNGATAVFNTGIFQYQQALANMQLQQHTAFLPPGSILCMTPATSVVPMVHGATPATVSAATTSATSVPFAATATANQIPIISAEHLTSHKYVTQMSpecifications
| MBNL1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length recombinant MBNL1. | |
| MBNL1 | |
| DKFZp686P06174/EXP/EXP35/EXP40/EXP42/KIAA0428/MBNL | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| BC043493 | |
| MBNL1 | |
| MBNL1 (AAH43493, 1 a.a. ∽ 382 a.a) full-length recombinant protein with GST tag. | |
| RUO | |
| 4154 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction