missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-MBL-2, Clone: 14D12, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 5 publications
Brand: Novus Biologicals NBP1-60128-0.05MG
This item is not returnable.
View return policy
Description
MBL-2 Monoclonal specifically detects MBL-2 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunoassay.
Specifications
| MBL-2 | |
| Monoclonal | |
| 0.1 mg/mL | |
| Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunoassay 1 ug/ml | |
| L-MBP, mannose-binding protein C, mannose-binding lectin (protein C) 2, MBL, MBL-C, MBP-C | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 17195 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| 14D12 | |
| Unconjugated | |
| 0.2 um filtered solution in PBS and 0.1% BSA with 0.02% Sodium Azide | |
| Mbl2 | |
| Mouse MBL-C | |
| 0.05 mg | |
| Primary | |
| Mouse | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction