missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mast cell tryptase, Mouse, Clone: AA1, Abnova™
Mouse monoclonal antibody raised against native Mast cell tryptase.
Brand: Abnova MAB5339.200ug
This item is not returnable.
View return policy
Specifications
| Mast cell tryptase | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:10000) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 200 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot | |
| AA1 | |
| Mouse monoclonal antibody raised against native Mast cell tryptase. | |
| In PBS (0.09% sodium azide) | |
| Native purified Mast cell tryptase from human lung tissue. | |
| RUO | |
| This antibody recognizes human mast cell tryptase, both alpha and beta isoforms. It is an excellent marker for mast cells, and does not bind to any other cell type in immunohistology. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction