Learn More
MAP3K7IP2, Mouse, Clone: 3B5, Abnova™
Mouse monoclonal antibody raised against recombinant MAP3K7IP2.
Brand: Abnova MAB10330.100uL
Description
The protein encoded by this gene is an activator of MAP3K7/TAK1, which is required for for the IL-1 induced activation of nuclear factor kappaB and MAPK8/JNK. This protein forms a kinase complex with TRAF6, MAP3K7 and TAB1, thus serves as an adaptor linking MAP3K7 and TRAF6. This protein, TAB1, and MAP3K7 also participate in the signal transduction induced by TNFSF11/RANKl through the activation of the receptor activator of NF-kappB (TNFRSF11A/RANK), which may regulate the development and function of osteoclasts. [provided by RefSeq
Specifications
| MAP3K7IP2 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Flow cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. | |
| MAP3K7IP2 | |
| MAP3K7IP2 | |
| Recombinant protein corresponding to human MAP3K7IP2. | |
| RUO | |
| 23118 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Flow Cytometry, Western Blot | |
| 3B5 | |
| Mouse monoclonal antibody raised against recombinant MAP3K7IP2. | |
| In ascites (0.03% sodium azide) | |
| FLJ21885/KIAA0733/TAB2 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.