Learn More
MAML1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant MAML1.
Brand: Abnova H00009794-A01.50uL
Description
This protein is the human homolog of mastermind, a Drosophila protein that plays a role in the Notch signaling pathway involved in cell-fate determination. There is in vitro evidence that the human homolog forms a complex with the intracellular portion of human Notch receptors and can increase expression of a Notch-induced gene. This evidence supports its proposed function as a transcriptional co-activator in the Notch signaling pathway. [provided by RefSeq
Sequence: AEQEKQQFQRHLTRPPPQYQDPTQGSFPQQVGQFTGSSAAVPGMNTLGPSNSSCPRVFPQAGNLMPMGPGHASVSSLPTNSGQQDRGVAQFPGSQNMPQSSpecifications
| MAML1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant MAML1. | |
| MAML1 | |
| KIAA0200/Mam-1/Mam1 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_014757 | |
| MAML1 | |
| MAML1 (NP_055572, 655 a.a. ∽ 754 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 9794 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only