Learn More
MALT1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant MALT1.
Brand: Abnova H00010892-A01.50uL
Description
This gene has been found to be recurrently rearranged in chromosomal translocation with two other genes - baculoviral IAP repeat-containing protein 3 (also known as apoptosis inhibitor 2) and immunoglobulin heavy chain locus - in mucosa-associated lymphoid tissue lymphomas. The protein encoded by this gene may play a role in NF-kappaB activation. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq
Sequence: EDTVEDKQEVNVGKPLIAKLDMHRGLGRKTCFQTCLMSNGPYQSSAATSGGAGHYHSLQDPFHGVYHSHPGNPSNVTPADSCHCSRTPDAFISSFAHHASSpecifications
| MALT1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant MALT1. | |
| MALT1 | |
| DKFZp434L132/MLT/MLT1 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| BC030143 | |
| MALT1 | |
| MALT1 (AAH30143, 685 a.a. ∽ 784 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 10892 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only