Learn More
MALT1, Mouse, Clone: MT1/410, Abnova™
Mouse monoclonal antibody raised against MALT1.
Brand: Abnova MAB11232.100ug
Description
This gene has been found to be recurrently rearranged in chromosomal translocation with two other genes - baculoviral IAP repeat-containing protein 3 (also known as apoptosis inhibitor 2) and immunoglobulin heavy chain locus - in mucosa-associated lymphoid tissue lymphomas. The protein encoded by this gene may play a role in NF-kappaB activation. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq
Specifications
| MALT1 | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1:400) The optimal working dilution should be determined by the end user. | |
| MALT1 | |
| MALT1 | |
| Recombinant protein corresponding to human MALT1. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| MT1/410 | |
| Mouse monoclonal antibody raised against MALT1. | |
| In PBS (0.05% BSA, 0.05% sodium azide) | |
| DKFZp434L132/MLT/MLT1 | |
| Mouse | |
| Protein G affinity chromatography | |
| RUO | |
| 10892 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.