Learn More
LY96 (B03P), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial human LY96 protein.
Brand: Abnova H00023643-B03P.50ug
Description
The MD-2 protein appears to associate with toll-like receptor 4 on the cell surface and confers responsiveness to lipopolysaccyaride (LPS), thus providing a link between the receptor and LPS signaling. [provided by RefSeq
Sequence: QKQYWVCNSSDASISYTYCDKMQYPISINVNPCIELKGSKGLLHIFYIPRRDLKQLYFNLYITVNTMNLPKRKEVICRGSDDDYSFCRALKGETVNTTISFSFKGIKFSKGKYKCVVEAISGSPEEMLFCLEFVILHQPNSNSpecifications
| LY96 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial human LY96 protein. | |
| LY96 | |
| MD-2/MD2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 23643 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC020690 | |
| LY96 | |
| LY96 (AAH20690,19 a.a. ∽ 160 a.a) partial human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only