Learn More
LY9, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant LY9.
Brand: Abnova H00004063-A01.50uL
Description
LY9 belongs to the SLAM family of immunomodulatory receptors (see SLAMF1; MIM 603492) and interacts with the adaptor molecule SAP (SH2D1A; MIM 300490) (Graham et al., 2006 [PubMed 16365421]).[supplied by OMIM
Sequence: EPQVTMKSVKVSENFSCNITLMCSVKGAEKSVLYSWTPREPHASESNGGSILTVSRTPCDPDLPYICTAQNPVSQRSSLPVHVGQFCTDPGASRGGTTSpecifications
| LY9 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant LY9. | |
| LY9 | |
| CD229/SLAMF3/hly9/mLY9 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_002348 | |
| LY9 | |
| LY9 (NP_002339, 156 a.a. ∽ 253 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 4063 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only