missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LPA, Mouse, Clone: 8F6A9, 8H5C5, Abnova™
Mouse monoclonal antibody raised against partial recombinant LPA.
Brand: Abnova MAB10563.100uL
This item is not returnable.
View return policy
Specifications
| LPA | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user. | |
| LPA | |
| LPA | |
| Recombinant protein corresponding to amino acids 4330-4521 of human LPA. | |
| RUO | |
| 4018 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunohistochemistry, Western Blot | |
| 8F6A9, 8H5C5 | |
| Mouse monoclonal antibody raised against partial recombinant LPA. | |
| In ascites (0.03% sodium azide) | |
| AK38/APOA/LP | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction