missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMTK3, Mouse, Clone: 2H6, Abnova™
Mouse monoclonal antibody raised against a partial recombinant LMTK3.
Brand: Abnova H00114783-M02A.200uL
This item is not returnable.
View return policy
Description
Sequence: VLVNGGLTPPKSEDKVSENGGLRFPRNTERPPETGPWRAPGPWEKTPESWGPAPTIGEPAPETSLERAPAPSAVVSSRNGGETAPGPLGPAPKNGTLEPGSpecifications
| LMTK3 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| XM_055866 | |
| LMTK3 | |
| LMTK3 (XP_055866,1151 a.a. ∽ 1250 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 114783 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 2H6 | |
| Mouse monoclonal antibody raised against a partial recombinant LMTK3. | |
| LMTK3 | |
| KIAA1883/LMR3/TYKLM3 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction