Learn More
LIPA, Mouse, Clone: 9G7F12, Abnova™
Mouse monoclonal antibody raised against partial recombinant LIPA.
Brand: Abnova MAB5527.100ug
Description
This gene encodes lipase A, the lysosomal acid lipase (also known as cholesterol ester hydrolase). This enzyme functions in the lysosome to catalyze the hydrolysis of cholesteryl esters and triglycerides. Mutations in this gene can result in Wolman disease and cholesteryl ester storage disease. Alternatively spliced transcript variants encoding the same protein have been found for this gene. [provided by RefSeq
Specifications
| LIPA | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1:200-1:1000) ELISA (1:10000) The optimal working dilution should be determined by the end user. | |
| LIPA | |
| LIPA | |
| Recombinant protein corresponding to human LIPA. | |
| RUO | |
| 3988 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Western Blot | |
| 9G7F12 | |
| Mouse monoclonal antibody raised against partial recombinant LIPA. | |
| In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
| CESD/LAL | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.