Learn More
LHX2, Mouse, Clone: 6G2, Abnova™
Mouse monoclonal antibody raised against partial recombinant LHX2.
Brand: Abnova MAB10789.100uL
Description
This gene encodes a protein belonging to a large protein family, members of which carry the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator. The protein can recapitulate or rescue phenotypes in Drosophila caused by a related protein, suggesting conservation of function during evolution. [provided by RefSeq
Specifications
| LHX2 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunofluorescence (1:200-1:1000) The optimal working dilution should be determined by the end user. | |
| LHX2 | |
| LHX2 | |
| Recombinant protein corresponding to human LHX2. | |
| RUO | |
| 9355 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunofluorescence, Western Blot | |
| 6G2 | |
| Mouse monoclonal antibody raised against partial recombinant LHX2. | |
| In ascites (0.03% sodium azide) | |
| LH2/MGC138390/hLhx2 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.