Learn More
LGR4, Mouse, Clone: 12H6, Abnova™
Mouse monoclonal antibody raised against full-length recombinant LGR4.
Brand: Abnova MAB6679.50ug
Description
G protein-coupled receptors (GPCRs) play key roles in a variety of physiologic functions. Members of the leucine-rich GPCR (LGR) family, such as GPR48, have multiple N-terminal leucine-rich repeats (LRRs) and a 7-transmembrane domain (Weng et al., 2008 [PubMed 18424556]).[supplied by OMIM
Specifications
| LGR4 | |
| Monoclonal | |
| Unconjugated | |
| Flow Cyt (1-5 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| LGR4 | |
| LGR4 | |
| Recombinant protein corresponding to full length human LGR4. | |
| RUO | |
| 55366 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Flow Cytometry, Western Blot | |
| 12H6 | |
| Mouse monoclonal antibody raised against full length recombinant LGR4. | |
| In PBS (0.1% proclin, 2% Block Ace) | |
| GPR48 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.