Learn More
LAT2, Mouse, PE, Clone: NAP-07, Abnova™
Mouse monoclonal antibody raised against partial recombinant LAT2.
Brand: Abnova MAB4522.100ug
Description
This gene is one of the contiguous genes at 7q11.23 commonly deleted in Williams syndrome, a multisystem developmental disorder. This gene consists of at least 14 exons, and its alternative splicing generates 3 transcript variants, all encoding the same protein. [provided by RefSeq
Specifications
| LAT2 | |
| Monoclonal | |
| 0.1 mg/mL | |
| Mouse monoclonal antibody raised against partial recombinant LAT2. | |
| In PBS (0.2% BSA, 0.09% sodium azide) | |
| HSPC046/LAB/NTAL/WBSCR15/WBSCR5/WSCR5 | |
| Mouse | |
| 100 μg | |
| Primary | |
| This antibody reacts with Non-T cell activation linker (NTAL), also known as LAB (linker of activated B cells), a 25-30 KDa transmembrane adaptor protein present in membrane microdomains (rafts) of B lymphocytes, NK cells and myeloid cells. | |
| Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot | |
| NAP-07 | |
| PE | |
| Flow Cytometry (1:30) The optimal working dilution should be determined by the end user. | |
| LAT2 | |
| LAT2 | |
| Recombinant protein corresponding to partial human LAT2. | |
| RUO | |
| 7462 | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.