missing translation for 'onlineSavingsMsg'
Learn More

Laminin, Mouse, Clone: MEC5, Abnova™

Product Code. 16065726
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16065726 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16065726 Supplier Abnova Supplier No. MAB2520.1mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against native Laminin.

Specifications

Antigen Laminin
Applications Immunohistochemistry (Frozen)
Classification Monoclonal
Clone MEC5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native Laminin.
Dilution The optimal working dilution should be determined by the end user.
Formulation In tissue culture supernatant (0.05% sodium azide)
Host Species Mouse
Immunogen Native purified rat Laminin.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This rat monoclonal antibody is directed against the P1 fragment of laminin. They are species-independent as it has been shown that they bind to laminin of rat, mouse and humans.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.