missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT6, Mouse, Clone: Ks6.KA12, Abnova™
Mouse monoclonal antibody raised against native KRT6.
Brand: Abnova MAB5176.1mL
This item is not returnable.
View return policy
Specifications
| KRT6 | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (1:10) The optimal working dilution should be determined by the end user. | |
| CK6A/CK6C/CK6D/K6A/K6C/K6D/KRT6C/KRT6D | |
| Mouse | |
| 1 mL | |
| Primary | |
| This antibody represents an excellent marker for non-keratinized squamous epithelia and proliferating cells of epidermis (e.g. within psoriatic lesions). It is suitable for discrimination of keratinizing and non-keratinizing squamous cell carcinoma versus e.g. poorly differentiated adenocarcinoma. Polypeptide reacting: Mr 56000 polypeptide (cytokeratin 6) of human squamous epithelia. | |
| Store at 4°C. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot | |
| Ks6.KA12 | |
| Mouse monoclonal antibody raised against native KRT6. | |
| In Tris, pH 7.4 (0.5% BSA, 0.09% sodium azide) | |
| KRT6A | |
| Native purified human KRT6. | |
| RUO | |
| 3853/3854/286887 | |
| Human, Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction