missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Klrb1c, Mouse, PE, Clone: PK136, Abnova™
Mouse monoclonal antibody raised against native Klrb1c.
Brand: Abnova MAB5777.100ug
This item is not returnable.
View return policy
Specifications
| Klrb1c | |
| Monoclonal | |
| PE | |
| Flow Cytometry (0.5 ug/106 cells) The optimal working dilution should be determined by the end user. | |
| Klrb1c | |
| Klrb1c | |
| Native purified Klrb1c from CE mouse spleen and bone marrow cells enriched for NK-1+ cells. | |
| RUO | |
| 17059 | |
| Mouse | |
| Liquid |
| Flow Cytometry, Immunoprecipitation | |
| PK136 | |
| Mouse monoclonal antibody raised against native Klrb1c. | |
| In PBS (0.09% sodium azide) | |
| AI462337/CD161/Ly-59/Ly55c/Ly59/NK-RP1/NK1.1/NKRP1/NKRP140/Nk-1/Nk-1.2/Nk1/Nk1.2/Nkrp1c | |
| Mouse | |
| 100 μg | |
| Primary | |
| Mouse NK1.1 alloantigen, Mr 80-85 KDa (39 KDa under reducing conditions). | |
| Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction