Learn More
KLK3 Mouse anti-Human, Monoclonal Antibody, Abnova™
Mouse monoclonal antibody raised against native KLK3.
Brand: Abnova MAB1403.1mg
Description
Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. Its protein product is a protease present in seminal plasma. It is thought to function normally in the liquefaction of seminal coagulum, presumably by hydrolysis of the high molecular mass seminal vesicle protein. Serum level of this protein, called PSA in the clinical setting, is useful in the diagnosis and monitoring of prostatic carcinoma. Alternate splicing of this gene generates several transcript variants encoding different isoforms. [provided by RefSeq
Specifications
| KLK3 | |
| Monoclonal | |
| Mouse monoclonal antibody raised against native KLK3. | |
| In 15mM KPO4 buffer, 0.85% NaCl, pH 7.4 (0.09% sodium azide) | |
| APS/KLK2A1/PSA/hK3 | |
| Mouse | |
| 1 mg | |
| Primary | |
| This antibody is specific to KLK3. No cross-reactivity with human albumin, AFP, CEA, PAP, CA 125, CA 15-3, or CA 19-9. | |
| Liquid |
| ELISA | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| KLK3 | |
| KLK3 | |
| Native purified human KLK3. | |
| RUO | |
| 354 | |
| Store at 4°C. | |
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.