missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KDM5B, Mouse, Clone: 1G10, Abnova™
Mouse monoclonal antibody raised against a partial recombinant KDM5B.
Brand: Abnova H00010765-M02A.200uL
This item is not returnable.
View return policy
Description
Sequence: AKRMRAEAMNIKIEPEETTEARTHNLRRRMGCPTPKCENEKEMKSSIKQEPIERKDYIVENEKEKPKSRSKKATNAVDLYVCLLCGSGNSpecifications
| KDM5B | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_006618 | |
| KDM5B | |
| KDM5B (NP_006609,231 a.a. ∽ 319 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 10765 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 1G10 | |
| Mouse monoclonal antibody raised against a partial recombinant KDM5B. | |
| KDM5B | |
| FLJ10538/FLJ12459/FLJ12491/FLJ16281/FLJ23670/KDM5B/PLU-1/PUT1/RBBP2H1A | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction