missing translation for 'onlineSavingsMsg'
Learn More

KCNQ5, Mouse, Polyclonal Antibody, Abnova™

Product Code. 16138746
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16138746 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16138746 Supplier Abnova Supplier No. H00056479A01.50uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse polyclonal antibody raised against a partial recombinant KCNQ5.

This gene is a member of the KCNQ potassium channel gene family that is differentially expressed in subregions of the brain and in skeletal muscle. The protein encoded by this gene yields currents that activate slowly with depolarization and can form heteromeric channels with the protein encoded by the KCNQ3 gene. Currents expressed from this protein have voltage dependences and inhibitor sensitivities in common with M-currents. They are also inhibited by M1 muscarinic receptor activation. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: QNLIRSTEELNIQLSGSESSGSRGSQDFYPKWRESKLFITDEEVGPEETETDTFDAAPQPAREAAFASDSLRTGRSRSSQSICKAGESTDALSLPHVKLK

Specifications

Antigen KCNQ5
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant KCNQ5.
Formulation 50% glycerol
Gene KCNQ5
Gene Accession No. NM_019842
Gene Alias Kv7.5
Gene Symbols KCNQ5
Host Species Mouse
Immunogen KCNQ5 (NP_062816, 833 a.a. ∽ 932 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56479
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.