missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kcnab1, Mouse, Clone: S40-17, Abnova™
Mouse monoclonal antibody raised against full-length recombinant Kcnab1.
Brand: Abnova MAB8579.100ug
This item is not returnable.
View return policy
Specifications
| Kcnab1 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1 ug/mL) Immunohistochemistry (0.1-1.0 ug/mL) The optimal working dilution should be determined by the end user. | |
| Kcnab1 | |
| Kcnab1 | |
| Recombinant protein corresponding to full length of rat Kcnab1. | |
| RUO | |
| 29737 | |
| Human, Mouse, Rat | |
| Liquid |
| Immunocytochemistry, Western Blot | |
| S40-17 | |
| Mouse monoclonal antibody raised against full length recombinant Kcnab1. | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
| KvBeta3/MGC105463 | |
| Mouse | |
| 100 μg | |
| Primary | |
| This antibody is specific to Kcnab1. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction