missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kcna1, Mouse, Clone: S36-15, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of Kcna1.
Brand: Abnova MAB8577.100ug
This item is not returnable.
View return policy
Specifications
| Kcna1 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1 ug/mL) Immunohistochemistry (0.1-1.0 ug/mL) The optimal working dilution should be determined by the end user. | |
| Kcna1 | |
| Kcna1 | |
| A synthetic peptide corresponding to amino acids 191-208 of rat Kcna1. | |
| RUO | |
| 24520 | |
| Mouse, Rat | |
| Liquid |
| Immunocytochemistry, Western Blot | |
| S36-15 | |
| Mouse monoclonal antibody raised against synthetic peptide of Kcna1. | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
| Kcna/Kcpvd/Kv1.1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| This antibody is specific to Kcna1. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction