Learn More
KARS, Mouse, Clone: 8G12C1, Abnova™
Mouse monoclonal antibody raised against partial recombinant KARS.
Brand: Abnova MAB10375.100uL
Description
Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. Lysyl-tRNA synthetase is a homodimer localized to the cytoplasm which belongs to the class II family of tRNA synthetases. It has been shown to be a target of autoantibodies in the human autoimmune diseases, polymyositis or dermatomyositis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
| KARS | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user. | |
| KARS | |
| KARS | |
| Recombinant protein corresponding to amino acids 90-174 of human KARS. | |
| RUO | |
| 3735 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunohistochemistry, Western Blot | |
| 8G12C1 | |
| Mouse monoclonal antibody raised against partial recombinant KARS. | |
| In ascites (0.03% sodium azide) | |
| KARS2/KIAA0070/KRS | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.