missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JMY, Mouse, Clone: 5B6, Abnova™
Mouse monoclonal antibody raised against a partial recombinant JMY.
Brand: Abnova H00133746-M05A.200uL
This item is not returnable.
View return policy
Description
Sequence: SPMDEVLASLKRGSFHLKKVEQRTLPPFPDEDDSNNILAQIRKGVKLKKVQKDVLRESFTLLPDTDPLTRSIHEALRRIKEASPESEDEEEALPCTDWENSpecifications
| JMY | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_152405 | |
| JMY | |
| JMY (NP_689618,535 a.a. ∽ 634 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 133746 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 5B6 | |
| Mouse monoclonal antibody raised against a partial recombinant JMY. | |
| JMY | |
| FLJ37870/MGC163496 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction