missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITFG2, Mouse, Clone: 1A8, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant ITFG2.
Brand: Abnova H00055846-M01A.200uL
This item is not returnable.
View return policy
Description
Sequence: MRSVSYVQRVALEFSGSLFPHAICLGDVDNDTLNELVVGDTSGKVSVYKNDDSRPWLTCSCQGMLTCVGVGDVCNKGKNLLVAVSAEGWFHLFDLTPAKVLDASGHHETLIGEEQRPVFKQHIPANTKVMLISDIDGDGCRELVVGYTDRVVRAFRWEELGEGPEHLTGQLVSLKKWMLEGQVDSLSVTLGPLGLPELMVSQPGCAYAILLCTWKKDTGSPPASEGPTDGSRETPAARDVVLHQTSGRIHNKNVSTHLIGNIKQGHGTESSGSGLFALCTLDGTLKLMEEMEEADKLLWSVQVDHQLFALEKLDVTGNGHEEVVACAWDGQTYIIDHNRTVVRFQVDENIRAFCAGLYACKEGRNSPCLVYVTFNQKIYVYWEVQLERMESTNLVKLLETKPEYHSLLQELGVDPDDLPVTRALLHQTLYHPDQPPQCAPSSLQDPTSpecifications
| ITFG2 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| BC006552 | |
| ITFG2 | |
| ITFG2 (AAH06552,1 a.a. ∽ 447 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 55846 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 1A8 | |
| Mouse monoclonal antibody raised against a full-length recombinant ITFG2. | |
| ITFG2 | |
| MDS028 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction