missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IQGAP3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant IQGAP3.
Brand: Abnova H00128239-A01.50uL
This item is not returnable.
View return policy
Description
Sequence: DTNTAQRMALGVAAINQAIKEGKAAQTERVLRNPAVALRGVVPDCANGYQRALESAMAKKQRPADTAFWVQHDMKDGTAYYFHLQTFQGIWEQPPGCSpecifications
| IQGAP3 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant IQGAP3. | |
| IQGAP3 | |
| MGC10170/MGC10831/MGC1947 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_178229 | |
| IQGAP3 | |
| IQGAP3 (NP_839943, 602 a.a. ∽ 698 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 128239 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur