Learn More
INVS, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant INVS.
Brand: Abnova H00027130-A01.50uL
Description
This gene encodes a protein containing multiple ankyrin domains and two IQ calmodulin-binding domains. The encoded protein may function in renal tubular development and function, and in left-right axis determination. This protein interacts with nephrocystin and infers a connection between primary cilia function and left-right axis determination. A similar protein in mice interacts with calmodulin. Mutations in this gene have been associated with nephronophthisis type 2. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq
Sequence: MNKSENLLFAGSSLASQVHAAAVNGDKGALQRLIVGNSALKDKEDQFGRTPLMYCVLADRLDCADALLKAGADVNKTDHSQRTALHLAAQKSpecifications
| INVS | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant INVS. | |
| INVS | |
| INV/KIAA0573/MGC133080/MGC133081/NPH2/NPHP2 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_014425 | |
| INVS | |
| INVS (NP_055240, 1 a.a. ∽ 91 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 27130 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
For Research Use Only