missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza B virus nucleoprotein, Mouse, Clone: InB27, Abnova™
Mouse monoclonal antibody raised against influenza B virus nucleoprotein.
Brand: Abnova MAB3154.200ug
This item is not returnable.
View return policy
Specifications
| Influenza B virus nucleoprotein | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 200 μg | |
| Primary | |
| Virus | |
| Liquid |
| ELISA, Western Blot | |
| InB27 | |
| Mouse monoclonal antibody raised against influenza B virus nucleoprotein. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| Influenza virus type B. | |
| RUO | |
| This antibody detects with nucleoprotein of influenza virus type B. | |
| Store at 4°C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction