Learn More
IMP-2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant IMP-2.
Brand: Abnova H00010644-A01.50uL
Description
This gene encodes a member of the IGF-II mRNA-binding protein (IMP) family. The protein encoded by this gene contains several four KH domains and two RRM domains. It functions by binding to the 5' UTR of the insulin-like growth factor 2 (IGF2) mRNA and regulating IGF2 translation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]
Sequence: HGKIMEVDYSVSKKLRSRKIQIRNIPPHLQWEVLDGLLAQYGTVENVEQVNTDTETAVVNVTYATREEAKIAMEKLSGHQFENYSFKISYIPDEEVSSPSPPQRASpecifications
| IMP-2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant IMP-2. | |
| IGF2BP2 | |
| IMP-2/IMP2/VICKZ2/p62 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_006548 | |
| IGF2BP2 | |
| IMP-2 (NP_006539, 65 a.a. ∽ 169 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 10644 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only