Learn More
IMMP2L, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant IMMP2L.
Brand: Abnova H00083943-A01.50uL
Description
The mitochondrial inner membrane peptidase (IMP) complex generates mature, active proteins in the mitochondrial intermembrane space by proteolytically removing the mitochondrial targeting presequence of nuclear-encoded proteins. IMP1 (IMMP1L; MIM 612323) and IMP2 are the catalytic subunits of the IMP complex (Burri et al., 2005 [PubMed 15814844]).[supplied by OMIM
Sequence: DRVACVARVEGASMQPSLNPGGSQSSDVVLLNHWKVRNFEVHRGDIVSLVSPKNPEQKIIKRVIALEGDIVRSpecifications
| IMMP2L | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant IMMP2L. | |
| IMMP2L | |
| IMP2/IMP2-LIKE | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_032549 | |
| IMMP2L | |
| IMMP2L (NP_115938, 31 a.a. ∽ 102 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 83943 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only