Learn More
IL34, Mouse, Clone: 1D12, Abnova™
Mouse monoclonal antibody raised against partial recombinant IL34.
Brand: Abnova MAB10698.100uL
Description
Interleukin-34 is a cytokine that promotes the differentiation and viability of monocytes and macrophages through the colony-stimulating factor-1 receptor (CSF1R; MIM 164770) (Lin et al., 2008 [PubMed 18467591]).[supplied by OMIM
Specifications
| IL34 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunofluorescence (1:200-1:1000) Flow cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. | |
| IL34 | |
| IL34 | |
| Recombinant protein corresponding to human IL34. | |
| RUO | |
| 146433 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Flow Cytometry, Immunofluorescence, Western Blot | |
| 1D12 | |
| Mouse monoclonal antibody raised against partial recombinant IL34. | |
| In ascites (0.03% sodium azide) | |
| C16orf77/IL-34/MGC34647 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.