missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-IGF-I, Clone: M23, Azide Free, Novus Biologicals™
Mouse Monoclonal Antibody
Specifications
| Antigen | IGF-I |
|---|---|
| Clone | M23 |
| Concentration | 1.0 mg/mL |
| Immunogen | Purified human IGF-1 protein |
| Gene Accession No. | P05019 |
Description
IGF-I/IGF-1 Monoclonal antibody specifically detects IGF-I/IGF-1 in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| IGF-I | |
| 1.0 mg/mL | |
| P05019 | |
| 10mM PBS - BSA free with No Preservative | |
| This antibody is specific to Insulin-like Growth Factor (IGF-1) and shows minimal cross-reaction with IGF-11, Proinsulin, MSF, and Insulin. IGF-1 is a polypeptide growth factor with two isoforms that are produced by alternative splicing. Isoform 1 is also known as IGF-IB while isoform 2 is known as IGF-IA. IGF-1 stimulates the proliferation of a wide range of cell types including muscle, bone and cartilage tissue. It functions as an autocrine regulator of growth. Activation of IGF system has emerged as a key factor for tumor progression and resistance to apoptosis in many cancers like those of breast, thyroid and colon. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ | |
| Protein A purified | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| M23 | |
| Purified human IGF-1 protein | |
| 3479 | |
| Protein A purified | |
| Primary | |
| Mouse | |
| Unconjugated | |
| RUO |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title