missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Varicella zoster virus (gpI/IV), Mouse, Clone: SG1, Abnova™
Mouse monoclonal antibody raised against varicella zoster virus (gpI/IV) .
Brand: Abnova MAB6698.500ug
This item is not returnable.
View return policy
Specifications
| Varicella zoster virus (gpI/IV) | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 500 μg | |
| Primary | |
| Virus | |
| Liquid |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| SG1 | |
| Mouse monoclonal antibody raised against varicella zoster virus (gpI/IV) . | |
| In 20mM Na2HPO4, pH 9.0 | |
| Varicella-Zoster Virus Ellen strain from VZV-infected monkey kidney cells (BSC-1). | |
| RUO | |
| Reacts with precursor as well as with mature VZV glycoprotein I and glycoprotein IV. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction