missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Streptococcus pneumoniae, Mouse, Clone: XI64, Abnova™
Mouse monoclonal antibody raised against Streptococcus pneumoniae.
Brand: Abnova MAB8259.200ug
This item is not returnable.
View return policy
Specifications
| Streptococcus pneumoniae | |
| Monoclonal | |
| Mouse monoclonal antibody raised against Streptococcus pneumoniae. | |
| In PBS | |
| Native Streptococcus pneumoniae (strain R36A). | |
| RUO | |
| These antibodies specifically recognize pneumococcal surface protein A. | |
| Liquid |
| ELISA | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 200 μg | |
| Primary | |
| Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction