missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Streptococcus agalactiae, Mouse, Clone: Strep12-504.5, Abnova™
Mouse monoclonal antibody raised against Streptococcus agalactiae.
Brand: Abnova MAB8390.100ug
This item is not returnable.
View return policy
Specifications
| Streptococcus agalactiae | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Bacteria | |
| Liquid |
| ELISA | |
| Strep12-504.5 | |
| Mouse monoclonal antibody raised against Streptococcus agalactiae. | |
| In PBS, pH 7.4 | |
| Native Streptococcus agalactiae cells isolated from mastitic milk. | |
| RUO | |
| These antibodies recognize Strep. agalactiae and cross-react weakly with Strep. dysgalactiae. They do not crossreact with Strep. uberis, Strep. bovis, Strep. faecalis, Staph. aureus, Mycoplasma bovis, Mycoplasma californicum, Mycoplasma bovigenitalium, E. coli, or Corynebacterium bovis. | |
| Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction