missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Salmonella typhi, Mouse, Clone: AC04, Abnova™
Mouse monoclonal antibody raised against native Salmonella typhi.
Brand: Abnova MAB2443.100uL
This item is not returnable.
View return policy
Specifications
| Salmonella typhi | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μL | |
| Primary | |
| S. typhi | |
| Liquid |
| ELISA, Immunofluorescence | |
| AC04 | |
| Mouse monoclonal antibody raised against native Salmonella typhi. | |
| In PBS (0.09% sodium azide) | |
| Native purified Salmonella typhi. | |
| RUO | |
| This antibody is only reactive with S. typhi. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction