missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlamydia trachomatis LPS, Mouse, Clone: CL19-158.3.1, Abnova™
Mouse monoclonal antibody raised against Chlamydia trachomatis LPS.
Brand: Abnova MAB6165.100ug
This item is not returnable.
View return policy
Specifications
| Chlamydia trachomatis LPS | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Bacteria | |
| Liquid |
| ELISA, Immunofluorescence | |
| CL19-158.3.1 | |
| Mouse monoclonal antibody raised against Chlamydia trachomatis LPS. | |
| In PBS, pH 7.4 | |
| Chlamydia trachomatis elementary bodies, L2 serovar. | |
| RUO | |
| These antibodies recognize Chlamydia trachomatis serovars A, B, Ba, C, D, E, F, G, H, I, J, K, L1, L2, L3. | |
| Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction