Learn More
Mouse anti-Human Kappa Light Chain Secondary Antibody, FITC, Invitrogen™
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen MH10511
123.50 GBP valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Code : 50
Additional Details : Weight : 0.50000kg
Description
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility. This antibody recognizes free light chains as well as those that are bound to heavy chains.
Human Kappa Light Chain Monoclonal antibody specifically detects Human Kappa Light Chain in Human samples. It is validated for Flow Cytometry.Specifications
Human Kappa Light Chain, Mouse anti-Human Kappa Light Chain Secondary Antibody, FITC | |
Monoclonal | |
FITC | |
Mouse | |
500 μL | |
Secondary | |
4° C, store in dark | |
IgG3 |
Flow Cytometry | |
HP6062 | |
PBS with BSA and 0.1% sodium azide | |
purified | |
ASR | |
Human | |
Liquid |