missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgM Fc Secondary Antibody (CH2), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-468DL594
This item is not returnable.
View return policy
Description
IgM Fc Monoclonal antibody specifically detects IgM Fc in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Specifications
| IgM Fc | |
| Monoclonal | |
| DyLight 594 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
| CH2 | |
| 50mM Sodium Borate | |
| Purified human IgM. | |
| 0.1 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction